Anti-H3K9me1 (methylated) Rabbit Polyclonal Antibody

Supplier: Prosci
18-621
76762-760EA 588.55 CAD
76762-760
Anti-H3K9me1 (methylated) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: H3K9me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Other
Order Now

SPECIFICATIONS

Antigen symbol H3K9me1
Antigen name Histone cluster 3, H3
Environmentally Preferable
Antigen synonyms H3.4|H3F3A|H3FT|H3t|H3/g
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Other,Human,Rat,Mouse
Host Rabbit
Gene ID 8290
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
ChIP Yes
Modification Methylated
Immunogen A synthetic methylated peptide corresponding to residues surrounding K9 of human histone H3.
Form Liquid
Purification Affinity purification
Concentration Batch dependent
Storage buffer PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
Storage temperature Store at −20...+730 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR