Anti-H3K36me1 (methylated) Rabbit Polyclonal Antibody

Supplier: Prosci
18-627
76761-342EA 588.55 CAD
76761-342
Anti-H3K36me1 (methylated) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: H3K36me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR