Anti-MSH6 Rabbit Polyclonal Antibody

Supplier: Prosci
18-253
76760-126EA 588.55 CAD
76760-126
Anti-MSH6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MSH6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR