Anti-HuDreg-55 Recombinant Antibody

Supplier: Prosci
10-286
76756-726EA 782.1 CAD
76756-726
Anti-HuDreg-55 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa-lambda
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR