Anti-ADAM5 Rabbit Polyclonal Antibody

Supplier: Prosci
22-049
76755-650EA 662.02 CAD
76755-650
Anti-ADAM5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ADAM5
Antigen name ADAM metallopeptidase domain 5 (pseudogene)
Environmentally Preferable
Antigen synonyms a disintegrin and metalloproteinase domain 5|TMDCII|ADAM5P|tMDC II
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 255926
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant protein of human ADAM5.
Form Liquid
Purification Affinity purification
Concentration Batch dependent
Storage buffer PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
Storage temperature Store at −20...+730 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR