Anti-HuPRO-140 Recombinant Antibody

Supplier: Prosci
10-888
76752-838EA 782.1 CAD
76752-838
Anti-HuPRO-140 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR