Anti-CIAO1 Rabbit Polyclonal Antibody

Supplier: Prosci
22-888
76752-500EA 492.88 CAD
76752-500
Anti-CIAO1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CIAO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR