Anti-CMC-544 Recombinant Antibody

Supplier: Prosci
10-275
76751-840EA 782.1 CAD
76751-840
Anti-CMC-544 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR