Anti-YTHDF3 Rabbit Polyclonal Antibody

Supplier: Prosci
23-552
76750-394EA 492.88 CAD
76750-394
Anti-YTHDF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: YTHDF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR