Anti-SUB1 Rabbit Polyclonal Antibody

Supplier: Prosci
22-674
76749-510EA 492.88 CAD
76749-510
Anti-SUB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol SUB1
Antigen name SUB1 homolog (S. cerevisiae)
Environmentally Preferable
Antigen synonyms PC4|p14|P15
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID 10923
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human SUB1 (NP_006704.3).
Form Liquid
Purification Affinity purification
Concentration Batch dependent
Storage buffer PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
Storage temperature Store at −20...+730 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR