Anti-Lgals9 Recombinant Antibody [Clone: RG9-35]

Supplier: Prosci
24-626
76630-908EA 1086 CAD
76630-908
Anti-Lgals9 Recombinant Antibody [Clone: RG9-35]
Antibodies
Type: Primary
Antigen: lectin, galactose binding, soluble 9
Clonality: Recombinant
Clone: RG9-35
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol Lgals9
Environmentally Preferable
Antigen name lectin, galactose binding, soluble 9
Antigen synonyms ecalectin|tumor antigen HOM-HD-21|soluble 5|HUAT|Galectin-9|urate transporter/channel protein.|LGALS9A|galactose binding|galectin 9|Gal-9|lectin
Antibody type Primary
Clonality Recombinant
Conjugation Unconjugated
Clone RG9-35
Reactivity Mouse
Host
Gene ID 16859
Isotype IgG2a kappa
Flow cytometry Yes
Modification Unmodified
Immunogen This antibody was raised by immunising rats with recombinant mouse galectin-9.
Form Liquid
Cross adsorption No
Storage buffer PBS with 0.02% Proclin 300
Storage temperature Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
Shipping temperature Shipped on blue ice
Tested applications Blocking, functional assays

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR