Anti-Fluorescein (FITC) Recombinant Antibody [Clone: 4-4-20 (enhanced)]

Supplier: Prosci
24-071
76630-508EA 1086 CAD
76630-508
Anti-Fluorescein (FITC) Recombinant Antibody [Clone: 4-4-20 (enhanced)]
Antibodies
Type: Primary
Antigen: Fluorescein (FITC)
Clonality: Recombinant
Clone: 4-4-20 (enhanced)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Fab fragment kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR