Prostate Biopsy Kits, Mopec
Supplier: Mortech Manufacturing
BG670 BG671 BG672
76579-384EA 38.9 CAD
76579-384 76579-386 76579-388
Prostate Biopsy Kits, Mopec
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
The Mopec prostate biopsy kits come ready to use in a set of 8, 12, or 16 containers. Containers are 20 ml capacity, half-filled with 10% neutral buffered formalin, and feature a double-lock closure that guarantees the container’s seal is fully secured.
  • 20 ml containers are half-filled with 10% neutral buffered formalin
  • Feature a double-lock closure that guarantees the container’s seal is fully secured
  • Leak-resistant seal on containers are 95 kPa compliant
  • Contains specific instructions for use and a cardboard insert labeled for placement of the
More Product Information