Mini Section Dryer, Mopec
Supplier: Mortech Manufacturing
BK700 BK700-D BK700-R BK700-T
76579-296EA 7338.58 CAD
76579-296 76579-302 76579-300 76579-298
Mini Section Dryer, Mopec
Slide Warmers/Dryers
Mopec’s mini section dryer is designed to efficiently and rapidly blow temperature-controlled air through the base of the instrument to dry specimens without the risk of overheating. Through this process, drying times are dramatically reduced by 75% compared to conventional drying ovens.
  • Fan-assisted heating dramatically reduces drying time
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Timer unit (BK700-
More Product Information