Raised Dissection Table, Mortech
Supplier: Mortech Manufacturing
BC090
76549-614EA 1953.07 CAD
76549-614
Raised Dissection Table, Mortech
Furniture Tables and Workbenches
This raised dissection table is used to raise your dissection board above the body on your work surface.
  • All polished stainless steel construction
  • Heavy gauge material provides a very stable work surface
  • Tray creased toward center drain
  • Suitable for over the body placement
Drain has optional hose attachment to route flow to desired location.
Ordering information: Dissection Board sold separately.