Bone Chisel, U.S. Army Pattern, Mortech
Supplier: Mortech Manufacturing
AG002 AG002-H AG003 AG004 AG102 AG103 AG104
76549-190EA 70.52 CAD
76549-190 76549-192 76549-194 76549-196 76549-254 76549-256 76549-258
Bone Chisel, U.S. Army Pattern, Mortech
Chisels
Osteotomes, also called bone chisels, are used to score, cut, scrape, clean and sculpt the bone.
  • Stainless steel
  • U.S. Army pattern
Available in standard or premium grade with various edges.