Forceps, Hemostatic Rochester Pean, Mortech
Supplier: Mortech Manufacturing
AC005 AC006 AC007 AC008 AC009 AC010 AC011 AC012 AC013 AC014 AC015 AC016 AC105 AC106 AC107 AC108 AC109 AC110 AC111 AC112 AC113 AC114 AC115 AC116
76548-906EA 29.14 CAD
76548-906 76548-908 76548-910 76548-912 76548-914 76548-916 76548-918 76548-920 76548-922 76548-924 76548-926 76548-928 76548-958 76548-960 76548-962 76548-964 76548-966 76548-968 76548-970 76548-972 76548-974 76548-976 76548-978 76548-980
Forceps, Hemostatic Rochester Pean, Mortech
Forceps
Finger ring grip and adjustable interlocking cross arms for secure clamping. Optional curved end allows for added laboratory uses.
  • Stainless steel
  • Finger ring grip
  • Curved/straight, serrated tips
Available in standard or premium grade with straight or curved edges.