Scissors, Tungsten Carbide, Iris, Mortech
Supplier: Mortech Manufacturing
AA405 AA406
76548-528EA 581.93 CAD
76548-528 76548-530
Scissors, Tungsten Carbide, Iris, Mortech
Scissors
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Premium grade stainless steel
  • Sharp/sharp points
Available with straight or curved edges.
Order Now
SPECIFICATIONS
Environmentally Preferable