100-Place Plastic Slide Box, Mortech®
Supplier: Mortech Manufacturing
BH010
76498-596EA 99.87 CAD
76498-596
100-Place Plastic Slide Box, Mortech®
Microscope Slide Boxes
Storage for up to 100 standard 1×3" microscope slides.
  • Hinged lid and positive snap closure
  • 100 slide capacity
Provides protection for slides in numbered slots with printed index card mounted inside of lid.