Cryostat Mounting Disks, Mortech®
Supplier: Mortech Manufacturing
BG010 BG011 BG012 BG013
76498-568EA 108.88 CAD
76498-568 76498-570 76498-572 76498-574
Cryostat Mounting Disks, Mortech®
Microtomes and Microtome Assemblies Microtomes
A drop of mounting media is placed on the disk and the specimen to be cut is fixed in place.
  • Available in four sizes
  • Chrome-plated
Order Now
SPECIFICATIONS
Environmentally Preferable