BEA (Bile Esculin Agar) with Azide/CNA (Colistin and Nalidixic Acid) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
J66
76441-340PK 770.91 CAD
76441-340
BEA (Bile Esculin Agar) with Azide/CNA (Colistin and Nalidixic Acid) Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Bile esculin agar with azide / columbia agar with 5% sheep blood, colistin and nalidixic acid for the presumptive identifcation of group D streptococci including enterococci and gram positive bacteria.
This Columbia CNA agar is recommended for use as a selective growth medium for the isolation and differentiation of Gram-positive cocci from clinical and non-clinical specimens which contain mixed flora.