GelGreen® Agarose LE, Biotium
Supplier: Biotium
41030-50G 41030-5G
76414-952EA 501.48 CAD
76414-952 76414-954
GelGreen® Agarose LE, Biotium
Electrophoresis Gels Agarose and Acrylamide
Ultrapure molecular biology grade agarose pre-coated with GelGreen® Nucleic acid gel stain for convenient and safer preparation of precast gels.
  • Non-toxic GelGreen® DNA dye is already in the agarose
  • No need to handle concentrated dye solutions, for enhanced safety
  • Use with TAE or TBE for 0.8 to 2% gels
  • Low EEO, ultrapure molecular biology grade agarose
  • Compatible with blue light illuminators
GelGreen® Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelGreen® Nucleic acid gel stain. There is no need to handleMore Product Information