INS Mouse Monoclonal Antibody [clone: 1G11] (ALEXA FLUOR® 750)
Supplier: Bioss
BSM-0855M-A750
76121-176EA 1434.2 CAD
76121-176
INS Mouse Monoclonal Antibody [clone: 1G11] (ALEXA FLUOR® 750)
Antibodies
Insulin is a pancreatic hormone that regulates glucose and is involved in the synthesis of protein and fat. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver. Heterodimer of a B chain and an A chain linked by two disulfide bonds.Belongs to the insulin family. The insulin-link growth factors, IGF-I and IGF-II (also desinated somatomedin C and multiplication stimulating activator, respectvely), share approximatly 76% sequence identity and are 50% related to pro-insulin.IGF-IMore Product Information