Anti-ANKRD26 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-9755R-A750
76120-484EA 1157.52 CAD
76120-484
Anti-ANKRD26 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network. Mutations of ankyrin genes lead to severe genetic diseases such as fatal cardiac arrhythmias and hereditary spherocytosis. ANKRD26 (ankyrin repeat domain-containing protein 26) is a 1709 amino acid protein that contains five ANK repeats. Expressed at high level in many tissues, including brain, liver, kidney and heart, ANKRD26 may be phosphorylated upon DNA damage by Atm or ATR. ANKRD26 is also expressed in the arcuate and ventromedial nucleiMore Product Information