Anti-NUDC Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-7841R-A750
76119-886EA 1157.52 CAD
76119-886
Anti-NUDC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
This gene encodes a nuclear distribution protein that plays an essential role in mitosis and cytokinesis. The encoded protein is involved in spindle formation during mitosis and in microtubule organization during cytokinesis. Pseudogenes of this gene are found on chromosome 2.

Type: Primary
Antigen: NUDC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 271-331/331
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR