Anti-DCTN6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-7747R-A680
76119-660EA 1157.52 CAD
76119-660
Anti-DCTN6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
DCTN6 belongs to the pointed end complex of the dynactin shoulder complex tpgether with the DCTN4, DCTN5 and DCTN6 subunits and ACTR10. DCTN6 contains an RGD (Arg-Gly-Asp) motif in the N terminal region, which confers adhesive properties to macromolecular proteins like fibronectin. The exact function of this gene is not known.

Type: Primary
Antigen: DCTN6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-190/190
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR