Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-7011R-A680
76119-068EA 1157.52 CAD
76119-068
Anti-SDCCAG8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-420/713
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR