Anti-Neurotensin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-6631R-A680
76118-492EA 1192.25 CAD
76118-492
Anti-Neurotensin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Neurotensin may play an endocrine or paracrine role in the regulation of fat metabolism. It causes contraction of smooth muscle.

Type: Primary
Antigen: Neurotensin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-170/170
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR