Anti-TPST1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-6850R-A680
76117-360EA 1157.52 CAD
76117-360
Anti-TPST1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Catalyzes the O-sulfation of tyrosine residues within acidic motifs of polypeptides.

Type: Primary
Antigen: TPST1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-260/370
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR