Anti-Cathelicidin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-6433R-A750
76116-284EA 1157.52 CAD
76116-284
Anti-Cathelicidin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Cathelicidins are a family of antimicrobial proteins found in the peroxidase-negative granules of neutrophils. Along with the family of proteins known as defensins, cathelicidins participate in the first line of defense by preventing local infection and systemic invasion of microbes. FALL-39 precursor (FALL-39 peptide antibiotic, cationic anti-microbial protein, CAMP, CAP-18, HSD26) is a cathelicidin anti-microbial protein that contains the antibacterial peptide LL-37 (amino acids 134-170). In contrast to the defensins, which are cysteine-rich peptides that fold in -pleated sheets, LL-37 isMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Cathelicidin
Antigen synonyms CAP 18|Cathelin-related antimicrobial peptide|CATHL3|Cathelin-like protein|mcramp|Cramp|CAP18|CAP-18|FALL 39 peptide antibiotic|LL37|Peptide antibiotic, PR-39, porcine, homolog of|CRAMP, mouse, homolog of|CLP|hCAP 18|Cationic antimicrobial protein CAP 18|FALL-39 peptide antibiotic|SOB3|18 kDa cationic antimicrobial protein|Cationic antimicrobial protein, 18-KD|hCAP18|Cnlp|CAMP|HSD26|hCAP-18|Antibacterial protein FALL-39|Cathelicidin antimicrobial peptide precursor|FALL 39|cathelicidin antimicrobial peptide|LL 37|MCLP|Antibacterial protein LL-37|CAMP_HUMAN|FALL39
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 750
Reactivity
Host Rabbit
Gene ID 820
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 101-170/170
Immunogen KLH conjugated synthetic peptide derived from human Calicidin
Form Liquid
Molecular weight 19 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
Storage temperature Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR