Anti-HSP40 homolog Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-6002R-A680
76116-146EA 1157.52 CAD
76116-146
Anti-HSP40 homolog Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
DNAJB4 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family, see MIM 608375.[supplied by OMIM, Mar 2008]

Type: Primary
Antigen: HSP40 homolog
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/337
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR