Anti-ALAS-E Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-9516R-A680
76110-726EA 1157.52 CAD
76110-726
Anti-ALAS-E Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
5-aminolevulinate synthase 1 (ALAS-H) and 2 (ALAS-E) are two isoforms of ALAS, an enzyme catalyzing the first step of the heme biosynthetic pathway in mammals. The erythroid-specific isoenzyme, ALAS-E, regulates the first step of hematopoietic cell differentiation and iron metabolism in the liver. ALAS-H is a housekeeping protein which mediates synthesis of early heme in the mitochondria of most cells. Succinyl CoA associates with ALAS-E in protein conformation change and translocation of ALAS-E into the mitochondria and does not interact with ALAS-H. The ALAS-E 5'-flanking region containsMore Product Information