Anti-MAP-9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-9313R-A680
76110-422EA 1157.52 CAD
76110-422
Anti-MAP-9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Microtubules, the primary component of the cytoskeletal network, interact with proteins called microtubule-associated proteins (MAPs).MAP9 is a microtubule-associated protein required for spindle function, mitotic progression, and cytokinesis.The microtubule-associated proteins can be divided into two groups, structural and dynamic. The MAP proteins function to stimulate tubulin assembly, enhance microtubule stability, influence the spatial distribution of microtubules within cells and utilize microtubule polarity to translocate cellular components. MAP-9 (microtubule-associated protein 9),More Product Information