Anti-DIP2A/B/C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-9312R-A750
76110-420EA 1157.52 CAD
76110-420
Anti-DIP2A/B/C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
DIP2A, also known as DIP2, is a 1571 amino acid nuclear protein. It is one of three human homologs (DIP2A, DIP2B and DIP2C) of the Drosophila dip2 (disconnected-interacting protein 2) protein. In Drosophila, dip2 interacts with disco, a protein required for neuronal connections in the visual systems of larvae and adults. The closest vertebrate homologs to disco are the basonuclin genes. In mice, DIP2 homologs show restricted expression to the brain. This suggests that, similar to the function of Drosophila dip2, vertebrate DIP2 homologs may play a role in the development of the nervousMore Product Information