Anti-HAGHL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-8414R-A680
76109-094EA 1192.27 CAD
76109-094
Anti-HAGHL Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The hydroxyacylglutathione hydrolase-like protein (HAGHL) is a 290 amino acid protein that belongs to the glyoxalase II family. HAGHL binds two zinc ions per subunit and acts as a hydrolase on ester bonds. The gene encoding HAGHL maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The rare disorder Rubinstein-Taybi syndrome is associated with chromosome 16, as is Crohn's disease, a gastrointestinal inflammatory condition that may involve the NOD2 gene.More Product Information