Anti-KLHL14 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-8056R-A750
76108-168EA 1157.52 CAD
76108-168
Anti-KLHL14 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
KLHL14 is a member of the KLHL family. The kelch motif was initially discovered in Kelch. In this protein there are six copies of the motif. It has been shown for one member that it is related to Galactose Oxidase for which a structure has been solved. Two isoforms of this protein exist - 70kDa and 43kDa.

Type: Primary
Antigen: KLHL14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 256-370/628
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR