Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-9509R-A750
76107-846EA 1157.52 CAD
76107-846
Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Leucine-rich repeats (LRRs) are 20-29 amino acid motifs that mediate protein-protein interactions. The primary function of these motifs is to provide a versatile structural framework for the formation of these protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervous system development. Several human diseases are associated with mutation in the genes encoding LRR-containing proteins. The leucine-rich repeat-containing protein 3B (LRRC3B), also designated LRP15, is a 259 amino acid protein that contains 3More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name LRP15
Antigen synonyms LRC3B_HUMAN|LRRC3B|Leucine rich repeat containing 3B|MGC102927|Leucine-rich repeat-containing protein 3B|LRP 15|Leucine-rich repeat protein LRP15|LRP-15
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 750
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 116135
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 181-259/259
Immunogen KLH conjugated synthetic peptide derived from human LRP15
Form Liquid
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR