Anti-T20 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Supplier: Bioss

BS-15561R-A750
76100-312EA 1190.99 CAD
76100-312
Anti-T20 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antibodies
IFT20 is a gene encodes a intraflagellar transport protein important for intracellular transport. The encoded protein forms part of a complex involved in trafficking of proteins from the Golgi body, including recycling of immune signalling components. This gene is part of a complex set of sense-antisense loci that may be co-regulated. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 14.

Type: Primary
Antigen: T20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR