Anti-CCL26 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Supplier: Bioss

BS-15512R-A680
76100-134EA 1190.99 CAD
76100-134
Anti-CCL26 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antibodies
CCL26 is a recently discovered CC chemokine. It binds to the CCR3 receptor and is highly chemotactic for eosinophils and basophils.

Type: Primary
Antigen: CCL26
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-94/94
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR