Anti-CD33 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-1514R-A750
76098-900EA 1190.99 CAD
76098-900
Anti-CD33 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,6-linked sialic acid (By similarity). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface.

Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/334
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR