Anti-S100B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-2015R-A680
76098-522EA 1190.99 CAD
76098-522
Anti-S100B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Weakly binds calcium but binds zinc very tightly-distinct binding sites with different affinities exist for both ions on each monomer. Physiological concentrations of potassium ion antagonize the binding of both divalent cations, especially affecting high-affinity calcium-binding sites. Binds to and initiates the activation of STK38 by releasing autoinhibitory intramolecular interactions within the kinase. Interaction with AGER after myocardial infarction may play a role in myocyte apoptosis by activating ERK1/2 and p53/TP53 signaling. Could assist ATAD3A cytoplasmic processing, preventingMore Product Information