Anti-Acetyl Histone H4K8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-4016R-A680
76085-608EA 1191 CAD
76085-608
Anti-Acetyl Histone H4K8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails; instead, they contain aMore Product Information