Anti-SDHD Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-3971R-A680
76085-488EA 1190.99 CAD
76085-488
Anti-SDHD Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Type: Primary
Antigen: SDHD
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-159/159
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name SDHD
Antigen synonyms Succinate-ubiquinone oxidoreductase cytochrome b small subunit|Succinate dehydrogenase complex subunit D|CII-4|SDHD|QPs3|CybS|Succinate-ubiquinone reductase membrane anchor subunit|Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial|SDH4
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Human,Rat
Host Rabbit
Gene ID 6392
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 81-159/159
Immunogen KLH conjugated synthetic peptide derived from human SDHD
Form Liquid
Molecular weight 11 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR