Anti-LTB4-R1/BLTR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-2654R-A750
76082-566EA 1190.99 CAD
76082-566
Anti-LTB4-R1/BLTR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Receptor for extracellular ATP > UTP and ADP. The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. May be the cardiac P2Y receptor involved in the regulation of cardiac muscle contraction through modulation of L-type calcium currents. Is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response.

Type: Primary
Antigen: LTB4-R1/BLTR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/352
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR