Anti-5-Lipoxygenase Ser271 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-3251R-A680
76082-164EA 1191 CAD
76082-164
Anti-5-Lipoxygenase Ser271 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a member of the lipoxygenase gene family and plays a dual role in the synthesis of leukotrienes from arachidonic acid. The encoded protein, which is expressed specifically in bone marrow-derived cells, catalyzes the conversion of arachidonic acid to 5(S)-hydroperoxy-6-trans-8,11,14-cis-eicosatetraenoic acid, and further to the allylic epoxide 5(S)-trans-7,9-trans-11,14-cis-eicosatetrenoic acid (leukotriene A4). Leukotrienes are important mediators of a number of inflammatory and allergic conditions. Mutations in the promoter region of this gene lead to a diminished responseMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name 5-Lipoxygenase Ser271
Antigen synonyms ALOX 5|Arachidonate 5 lipoxygenase|5LPG|LOG 5|MGC163204|ALOX5|LOG5|Arachidonic acid 5 lipoxygenase|LOX5_HUMAN|5-lipoxygenase|Leukotriene A4 synthase|5 LO
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Rat
Host Rabbit
Gene ID 240
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Phosphorylation (Ser271)
Immunogen KLH conjugated synthetic phosphopeptide derived from rat 5-Lipoxygenase around the phosphorylation site of Ser271
Form Liquid
Molecular weight 78 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR