Anti-SPHK1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-2652R-A680
76081-988EA 1190.99 CAD
76081-988
Anti-SPHK1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-sphingosine and to a lesser extent sphinganine, but not other lipids, such as D,L-threo-dihydrosphingosine, N,N-dimethylsphingosine, diacylglycerol, ceramide, or phosphatidylinositol.

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/384
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR