Anti-CYPA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-5912R-A680
76081-350EA 1163.04 CAD
76081-350
Anti-CYPA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Type: Primary
Antigen: CYPA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-165/165
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR