Anti-Calponin 2 + 3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-4902R-A750
76079-344EA 1190.99 CAD
76079-344
Anti-Calponin 2 + 3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Thin filament-associated protein that is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. The interaction of calponin with actin inhibits the actomyosin Mg-ATPase activity (By similarity).

Type: Primary
Antigen: Calponin 2 + 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/309
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR