Anti-Prostaglandin E Receptor EP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-4196R-A750
76077-964EA 1190.97 CAD
76077-964
Anti-Prostaglandin E Receptor EP2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. The subsequent raise in intracellular cAMP is responsible for the relaxing effect of this receptor on smooth muscle.

Type: Primary
Antigen: Prostaglandin E Receptor EP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 271-358/358
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR