Anti-TWF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-4169R-A680
76077-902EA 1190.99 CAD
76077-902
Anti-TWF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
PTK9 is an actin-binding protein involved in motile and morphological processes. It inhibits actin polymerization, likely by sequestering G-actin. By capping the barbed ends of filaments, it also regulates motility. PTK9 seems to play an important role in clathrin-mediated endocytosis and distribution of endocytic organelles.

Type: Primary
Antigen: TWF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 271-350/350
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR